(R)-(-)-1,2-Propanediol ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol
211835-1G
211835-25G
211835-5G
BT211835-1GEA
50
USD
BT211835-1G
BT211835-25G
BT211835-5G
(R)-(-)-1,2-Propanediol ≥98%
(R)-(-)-1,2-Propanediol
CAS: 4254-14-2; MDL No: MFCD00066248
Liquid; Linear Formula: CH3CH(OH)CH2OH; Molecular Formula: C3H8O2 ; MW: 76.09
Boiling Point: 186-188°; Flash point: 103°C (217°F)
Density (g/mL): 1.040 ; Refractive Index: 1.434; Optical Rotation: [α]20/D −16.5°, neat
Hygroscopic
- Optical rotation: [α]20/D −16.5°, neat
- Refractive index: 1.434
- Hygroscopic
- Liquid
Formula:
C₃H₈O₂ MW: 76.1 g/mol Boiling Pt: 186…188 °C Density: 1.040 g/mL Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00066248 CAS Number: 4254-14-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...