Magnesium(II) gluconate hydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: D-Gluconic acid hemimagnesium salt hydrate, Magnesium D-gluconate hydrate, Gluconic acid magnesium(II) salt hydrate, D-Gluconic acid magnesium salt hydrate

211820-100G 211820-500G
BT211820-100GEA 46 USD
BT211820-100G BT211820-500G
Magnesium(II) gluconate hydrate ≥98%
Magnesium(II) gluconate hydrate

CAS: 3632-91-5; EC No: 222-848-2; MDL No: MFCD00150971; RTECS: OM3480000 Solid; Molecular Formula: C12H22MgO14·xH2O; MW: 414.60 (anhydrous) Flash point: 100°C (212°F)


  • RTECS: OM3480000
  • Solid

Formula: C₆H₁₂O₇·xH₂O
MW: 414.60 g/mol (anhy)
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00150971
CAS Number: 3632-91-5
EINECS: 222-848-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR