Bis(dibenzylideneacetone)palladium

Supplier: BeanTown Chemical

Synonyms: Bis(dibenzylideneacetone)palladium(0)

211815-1G 211815-250MG 211815-5G
BT211815-1GEA 51 USD
BT211815-1G BT211815-250MG BT211815-5G
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium

CAS: 32005-36-0; MDL No: MFCD00051942 Powder; Molecular Formula: C34H28O2Pd; MW: 575.00 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR