Trithiocyanuric acid ≥95%
Supplier: BeanTown Chemical
211790-100G
211790-25G
211790-5G
BT211790-100GEA
542
USD
BT211790-100G
BT211790-25G
BT211790-5G
Trithiocyanuric acid ≥95%
Trithiocyanuric acid
CAS: 638-16-4; EC No: 211-322-8; MDL No: MFCD00006052; RTECS: XZ2830000
Powder; Molecular Formula: C3H3N3S3; MW: 177.27
Melting Point:
- RTECS: XZ2830000
- Powder
Formula:
C₃H₃N₃S₃ MW: 177.27 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00006052 CAS Number: 638-16-4 EINECS: 211-322-8 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...