Rhodium ≥99.5% (trace metals basis), wire Ø 1.0 mm

Supplier: BeanTown Chemical

211785-10CM 211785-2CM
BT211785-10CMEA 2303 USD
BT211785-10CM BT211785-2CM
Rhodium ≥99.5% (trace metals basis), wire Ø 1.0 mm
Rhodium

CAS: 7440-16-6; EC No: 231-125-0; MDL No: MFCD00011201 Solid; Molecular Formula: Rh; MW: 102.91 Melting Point: 1966°; Boiling Point: 3727° Density (g/mL): 12.41


  • Solid

Formula: Rh
Boiling Pt: 3727 °C
Melting Pt: 1966 °C
Density: 12.41 g/mL
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR