4'-Bromo-2'-hydroxyacetophenone ≥95%

Supplier: BeanTown Chemical

211705-1G 211705-250MG 211705-5G
BT211705-1GEA 94 USD
BT211705-1G BT211705-250MG BT211705-5G
4'-Bromo-2'-hydroxyacetophenone ≥95%
4'-Bromo-2'-hydroxyacetophenone

CAS: 30186-18-6 Solid; Molecular Formula: C8H7BrO2; MW: 215.04


  • Solid

Formula: C₈H₇BrO₂
MDL Number: MFCD03428533
CAS Number: 30186-18-6

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR