Cellulose, microcrystalline

Supplier: BeanTown Chemical

Synonyms: Microcrystallinecellulose

211700-10KG 211700-2.5KG
BT211700-10KGEA 477.47 USD
BT211700-10KG BT211700-2.5KG BT211700-500G
Cellulose, microcrystalline
Cellulose

CAS: 9004-34-6; EC No: 232-674-9; MDL No: MFCD00081512 Crystalline/Powder


  • Crystalline/Powder

Formula: (C₁₂H₂₂O₁₁)n
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR