2-Bromo-5-nitroanisole ≥98%

Supplier: BeanTown Chemical

211665-25G 211665-5G
BT211665-25GEA 240 USD
BT211665-25G BT211665-5G
2-Bromo-5-nitroanisole ≥98%
2-Bromo-5-nitroanisole

CAS: 77337-82-7; EC No: 278-669-5; MDL No: MFCD00041250 Solid; Molecular Formula: C7H6BrNO3; MW: 232.03


  • Solid

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD00041250
CAS Number: 77337-82-7
EINECS: 278-669-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR