Dimethyl (2-oxopropyl)phosphonate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Dimethyl acetylmethylphosphonate
211660-25G
211660-5G
BT211660-25GEA
324
USD
BT211660-25G
BT211660-5G
Dimethyl (2-oxopropyl)phosphonate ≥97%
Dimethyl (2-oxopropyl)phosphonate
CAS: 4202-14-6; EC No: 224-110-5; MDL No: MFCD00008769
Liquid; Molecular Formula: C5H11O4P; MW: 166.12
Boiling Point: 76-79°/3 mmHg; Flash point: 93°C (199°F)
Density (g/mL): 1.202; Refractive Index: 1.439
- Refractive index: 1.439
- Liquid
Formula:
C₅H₁₁O₄P Boiling Pt: 76…79 °C/3 mmHg Density: 1.202 g/mL Flash Pt: 93 °C (199 °F) |
MDL Number:
MFCD00008769 CAS Number: 4202-14-6 EINECS: 224-110-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...