Aluminium oxide, γ-phase ≥99.95%
Supplier: BeanTown Chemical
211655-100G
211655-25G
211655-500G
BT211655-100GEA
86
USD
BT211655-100G
BT211655-25G
BT211655-500G
Aluminium oxide, γ-phase ≥99.95%
Aluminium oxide, γ-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
3 Micron APS Powder, S.A. 80-120 m2/g; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 3 Micron APS Powder, S.A. 80-120 m²/g
Formula:
Al₂O₃ MW: 101.96 g/mol Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...