4-Hydroxybenzonitrile ≥98%
Supplier: BeanTown Chemical
Synonyms:
p-Cyanophenol, 4-Cyanophenol, p-Hydroxybenzonitrile
211645-250G
211645-50G
BT211645-250GEA
264
USD
BT211645-250G
BT211645-50G
4-Hydroxybenzonitrile ≥98%
4-Hydroxybenzonitrile
CAS: 767-00-0; EC No: 212-175-2; MDL No: MFCD00002312; RTECS: DI4375000
Crystalline; Molecular Formula: C7H5NO; MW: 119.12
Melting Point: 110-113°
- RTECS: DI4375000
- Crystalline
Formula:
C₇H₅NO MW: 119.12 g/mol Melting Pt: 110…113 °C |
MDL Number:
MFCD00002312 CAS Number: 767-00-0 EINECS: 212-175-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...