DL-Carnitine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
211620-100G
211620-25G
BT211620-100GEA
62
USD
BT211620-100G
BT211620-25G
DL-Carnitine hydrochloride ≥98%
DL-Carnitine hydrochloride
CAS: 461-05-2; EC No: 207-309-1; MDL No: MFCD00011904; RTECS: BP2979140
Crystalline; Molecular Formula: C7H17ClNO3; MW: 197.66
Hygroscopic
- RTECS: BP2979140
- Hygroscopic
- Crystalline
Formula:
C₇H₁₆ClNO₃ MW: 197.66 |
MDL Number:
MFCD00011904 CAS Number: 461-05-2 EINECS: 207-309-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...