2-Aminothiazole ≥97%
Supplier: BeanTown Chemical
Synonyms:
Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine
211610-250G
211610-50G
BT211610-250GEA
53
USD
BT211610-250G
BT211610-50G
2-Aminothiazole ≥97%
2-Aminothiazole
CAS: 96-50-4; EC No: 202-511-6; MDL No: MFCD00005325; RTECS: XJ2100000
Powder; Molecular Formula: C3H4N2S; MW: 100.14
Melting Point: 91-93°
- RTECS: XJ2100000
- Powder
Formula:
C₃H₄N₂S MW: 100.14 g/mol Melting Pt: 91…93 °C |
MDL Number:
MFCD00005325 CAS Number: 96-50-4 EINECS: 202-511-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...