Bismuth subsalicylate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Bismuth(III) subsalicylate, 2-Hydroxy-4H-benzo[d][1,3,2]dioxabismin-4-one
211585-1KG
211585-250G
BT211585-1KGEA
291
USD
BT211585-1KG
BT211585-250G
Bismuth subsalicylate ≥96%
Bismuth subsalicylate
CAS: 14882-18-9; EC No: 238-953-1; MDL No: MFCD00085368; RTECS: EB2985000
Powder, typically 2 micron max; Molecular Formula: C7H5BiO4 ; MW: 362.09
Melting Point:
- RTECS: EB2985000
- Powder, typically 2 micron max
Formula:
C₇H₅BiO₄ MW: 362.09 g/mol Melting Pt: >350 °C |
MDL Number:
MFCD00085368 CAS Number: 14882-18-9 EINECS: 238-953-1 |
Specification Test Results
Assay | 96% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...