n-Octadecane ≥99%
Supplier: BeanTown Chemical
CAS: 593-45-3; EC No: 209-790-3; MDL No: MFCD00009007
Wet Crystalline; Linear Formula: CH3(CH2)16CH3; Molecular Formula: C18H38; MW: 254.50
Melting Point: 26-29°; Boiling Point: 317°; Flash point: 166°C (331°F)
Density (g/mL): 0.777; Refractive Index: 1.439
- Refractive index: 1.439
- Crystalline
Formula:
H₃C(CH₂)₁₆CH₃ Boiling Pt: 317 °C Melting Pt: 26…29 °C Density: 0.777 g/mL Flash Pt: 166 °C (331 °F) |
MDL Number:
MFCD00009007 CAS Number: 593-45-3 EINECS: 209-790-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...