Silicon dioxide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
211575-1KG
211575-250G
BT211575-1KGEA
135
USD
BT211575-1KG
BT211575-250G
Silicon dioxide ≥99.5% (trace metals basis)
Silicon dioxide
CAS: 7631-86-9; EC No: 238-878-4; MDL No: MFCD00011232; RTECS: VV7330000
-325 Mesh Powder; Linear Formula: SiO2; MW: 60.08
Melting Point: 1610°; Boiling Point: 2230°
Density (g/mL): 2.2-2.6; Refractive Index: 1.544
- RTECS: VV7330000
- Refractive index: 1.544
- -325 Mesh Powder
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1610 °C Density: 2.2…2.6 g/mL |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 238-878-4 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...