Methyl palmitate ≥97%

Supplier: BeanTown Chemical

211570-100G 211570-500G
BT211570-100GEA 32.99 USD
BT211570-100G BT211570-500G
Methyl palmitate ≥97%
Methyl palmitate

CAS: 112-39-0; EC No: 203-966-3; MDL No: MFCD00008994 Solid; Linear Formula: CH3(CH2)14COOCH3; Molecular Formula: C17H34O2; MW: 270.45 Melting Point: 32-35°; Boiling Point: 185°/10 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.852; Refractive Index: 1.451


  • Refractive index: 1.451
  • Solid

Formula: C₁₇H₃₄O₂
Boiling Pt: 185 °C/10 mmHg
Melting Pt: 32…35 °C
Density: 0.852 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR