Yttrium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm

Supplier: BeanTown Chemical

211555-100MM 211555-25MM 211555-50MM
BT211555-100MMEA 989 USD
BT211555-100MM BT211555-25MM BT211555-50MM
Yttrium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm
Yttrium

CAS: 7440-65-5; EC No: 231-174-8; MDL No: MFCD00011468 Solid, cast rod, diameter is approximate; Molecular Formula: Y; MW: 88.91 Melting Point: 1522°; Boiling Point: 3338° Density (g/mL): 4.469


  • Solid, cast rod, diameter is approximate

Formula: Y
Boiling Pt: 3338 °C
Melting Pt: 1522 °C
Density: 4.469 g/mL
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR