1-Undecyne ≥97%
Supplier: BeanTown Chemical
211550-10G
211550-50G
BT211550-10GEA
91
USD
BT211550-10G
BT211550-50G
1-Undecyne ≥97%
1-Undecyne
CAS: 2243-98-3; EC No: 218-825-1; MDL No: MFCD00015063
Liquid; Linear Formula: CH3(CH2)8C≡CH; Molecular Formula: C11H20; MW: 152.28
Flash point: 65°C (149°F)
Density (g/mL): 0.770
- Liquid
Formula:
C₁₁H₂₀ MW: 152.28 g/mol Density: 0.770 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00015063 CAS Number: 2243-98-3 EINECS: 218-825-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...