3-(Trifluoromethyl)phenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: (3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane

211515-1G 211515-25G 211515-5G
BT211515-1GEA 50 USD
BT211515-1G BT211515-25G BT211515-5G
3-(Trifluoromethyl)phenylboronic acid ≥98%
3-(Trifluoromethyl)phenylboronic acid

CAS: 1423-26-3; MDL No: MFCD00151854 Powder; Molecular Formula: C7H6BF3O2; MW: 189.93 Melting Point: 163-166°


  • Powder

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 163…166 °C
MDL Number: MFCD00151854
CAS Number: 1423-26-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR