Potassium sulfate ≥99%
Supplier: BeanTown Chemical
211495-1KG
211495-5KG
BT211495-1KGEA
39.27
USD
BT211495-1KG
BT211495-5KG
Potassium sulfate ≥99%
Potassium sulphate
CAS: 7778-80-5; EC No: 231-915-5; MDL No: MFCD00011388; RTECS: TT5900000
Crystalline; Linear Formula: K2SO4; MW: 174.26
Melting Point: 1069°; Boiling Point: 1689°
Density (g/mL): 2.662
- RTECS: TT5900000
- Crystalline
Formula:
K₂SO₄ Boiling Pt: 1689 °C Melting Pt: 1069 °C Density: 2.662 |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...