Lanthanum(III) hydroxide ≥99.95%

Supplier: BeanTown Chemical

211475-250G 211475-50G
BT211475-250GEA 125 USD
BT211475-250G BT211475-50G
Lanthanum(III) hydroxide ≥99.95%
Lanthanum(III) hydroxide

CAS: 14507-19-8; EC No: 238-510-2; MDL No: MFCD00016160 Powder; Linear Formula: La(OH)3; MW: 189.93


  • Powder

Formula: H₃LaO₃
MW: 189.93
Density: 4.28
MDL Number: MFCD00016160
CAS Number: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8 III

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR