Aluminium sulfate, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

211470-5G 211470-25G 211470-100G
BT211470-5GEA 56.23 USD
BT211470-5G BT211470-100G BT211470-25G
Aluminium sulfate, anhydrous ≥99.99% (trace metals basis)
Aluminium sulphate

  • RTECS: BD1700000
  • Hygroscopic
  • Powder

Formula: Al₂O₁₂S₃
MW: 342.15 g/mol
Melting Pt: 770 °C
Density: 2.71 g/mL
MDL Number: MFCD00003423
CAS Number: 10043-01-3
EINECS: 233-135-0

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR