Iron(II) titanium oxide ≥99.5%
Supplier: BeanTown Chemical
211465-250G
211465-50G
BT211465-250GEA
671
USD
BT211465-250G
BT211465-50G
Iron(II) titanium oxide ≥99.5%
Iron(II) titanium oxide
CAS: 12022-71-8; EC No: 234-667-6; MDL No: MFCD00064704
-100 Mesh Powder; Linear Formula: FeTiO3; MW: 151.75
- -100 Mesh Powder
Formula:
FeTiO₃ MW: 151.75 g/mol |
MDL Number:
MFCD00064704 CAS Number: 12022-71-8 EINECS: 234-667-6 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...