Diisobutyl methylphosphonate ≥97%
Supplier: BeanTown Chemical
211445-1G
211445-5G
BT211445-1GEA
100
USD
BT211445-1G
BT211445-5G
Diisobutyl methylphosphonate ≥97%
Diisobutyl methylphosphonate
CAS: 7242-56-0; MDL No: MFCD00015638
Liquid; Molecular Formula: C9H21O3P; MW: 208.24
- Liquid
Formula:
C₉H₂₁O₃P MW: 208.24 g/mol |
MDL Number:
MFCD00015638 CAS Number: 7242-56-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...