2-Chlorobenzyl alcohol ≥99%

Supplier: BeanTown Chemical

Synonyms: (2-Chlorophenyl)methanol

211430-100G 211430-25G 211430-500G
BT211430-100GEA 176 USD
BT211430-100G BT211430-25G BT211430-500G
2-Chlorobenzyl alcohol ≥99%
2-Chlorobenzyl alcohol

CAS: 17849-38-6; EC No: 241-801-7; MDL No: MFCD00004604 Crystalline/Powder; Molecular Formula: C7H7ClO; MW: 142.58 Melting Point: 69-71°; Boiling Point: 227°


  • Crystalline/Powder

Formula: C₇H₇ClO
Boiling Pt: 227 °C
Melting Pt: 69…71 °C
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR