2-Nitrobenzaldehyde ≥98%
Supplier: BeanTown Chemical
211410-100G
211410-25G
211410-500G
BT211410-100GEA
149
USD
BT211410-100G
BT211410-25G
BT211410-500G
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde
CAS: 552-89-6; EC No: 209-025-3; MDL No: MFCD00007132; RTECS: CU7300000
Crystalline; Molecular Formula: C7H5NO3; MW: 151.12
Melting Point: 42-44°; Boiling Point: 153°/23 mmHg; Flash point:
- RTECS: CU7300000
- Air Sensitive, Light Sensitive
- Crystalline
Formula:
C₇H₅NO₃ MW: 151.12 g/mol Boiling Pt: 153 °C/23 mmHg Melting Pt: 42…44 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00007132 CAS Number: 552-89-6 EINECS: 209-025-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...