2,2'-Bithiophene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,2′-Dithienyl, 2,2′-Bithienyl

211405-1G 211405-25G 211405-5G
BT211405-1GEA 29 USD
BT211405-1G BT211405-25G BT211405-5G
2,2'-Bithiophene ≥98%
2,2'-Bithiophene

CAS: 492-97-7; EC No: 207-767-2; MDL No: MFCD00005414 Liquid; Molecular Formula: C8H6S2; MW: 166.26 Melting Point: 32-33°; Boiling Point: 260°; Flash point: 110°C (230°F) Light Sensitive


  • Light Sensitive
  • Liquid

Formula: C₈H₆S₂
MW: 166.27 g/mol
Boiling Pt: 260 °C
Melting Pt: 32…33 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR