3,4-Dihydroxybenzoic acid ≥97%
Supplier: BeanTown Chemical
211395-100G
211395-25G
211395-500G
BT211395-100GEA
164.13
USD
BT211395-100G
BT211395-25G
BT211395-500G
3,4-Dihydroxybenzoic acid ≥97%
3,4-Dihydroxybenzoic acid
CAS: 99-50-3; EC No: 202-760-0; MDL No: MFCD00002509; RTECS: UL0560000
Powder; Molecular Formula: C7H6O4; MW: 154.12
Melting Point: 197-200° (decomposes)
Air Sensitive
- RTECS: UL0560000
- Air Sensitive
- Powder
Formula:
C₇H₆O₄ Melting Pt: 197…200 °C (dec.) |
MDL Number:
MFCD00002509 CAS Number: 99-50-3 EINECS: 202-760-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...