Celite® standard super-cel, filter aid

Supplier: BeanTown Chemical

211380-2.5KG 211380-500G
BT211380-2.5KGEA 122 USD
BT211380-2.5KG BT211380-500G
Celite® standard super-cel, filter aid
Celite® standard super-cel, filter aid

CAS: 61790-53-2; EC No: 319-127-6; MDL No: MFCD00132803; RTECS: VV7311000 Powder, Lyophilized


  • RTECS: VV7311000
  • Powder, Lyophilized

MDL Number: MFCD00132803
CAS Number: 91053-39-3
EINECS: 319-127-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR