1-Octanesulphonic acid sodium salt monohydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate
211370-10G
211370-50G
BT211370-10GEA
62
USD
BT211370-10G
BT211370-50G
1-Octanesulphonic acid sodium salt monohydrate ≥99%
1-Octanesulphonic acid sodium salt monohydrate
CAS: 207596-29-0; EC No: 226-195-4; MDL No: MFCD00007544
Crystalline; Linear Formula: CH3(CH2)7SO3Na·H2O; Molecular Formula: C8H17NaO3S·H2O; MW: 234.29
Melting Point:
- Crystalline
Formula:
C₈H₁₇NaO₃S·1H₂O Melting Pt: >300 °C |
MDL Number:
MFCD00007544 CAS Number: 207596-29-0 EINECS: 226-195-4 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...