4-(Hydroxymethyl)phenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(Hydroxymethyl)benzeneboronic acid

211365-1G 211365-25G 211365-5G
BT211365-1GEA 39 USD
BT211365-1G BT211365-25G BT211365-5G
4-(Hydroxymethyl)phenylboronic acid ≥98%
4-(Hydroxymethyl)phenylboronic acid

CAS: 59016-93-2; MDL No: MFCD00792672 Solid; Molecular Formula: C7H9BO3; MW: 151.96 Melting Point: 251-256°


  • Solid

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 251…256 °C
MDL Number: MFCD00792672
CAS Number: 59016-93-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR