[Hydroxy(tosyloxy)iodo]benzene ≥97%

Supplier: BeanTown Chemical

211345-25G 211345-5G
BT211345-25GEA 114 USD
BT211345-25G BT211345-5G
[Hydroxy(tosyloxy)iodo]benzene ≥97%
[Hydroxy(tosyloxy)iodo]benzene

CAS: 27126-76-7; MDL No: MFCD00011547 Crystalline/Powder; Molecular Formula: C13H13IO4S; MW: 392.22 Melting Point: 131-137° Light Sensitive


  • Light Sensitive
  • Crystalline/Powder

Formula: C₁₃H₁₃IO₄S
Melting Pt: 131…137 °C
MDL Number: MFCD00011547
CAS Number: 27126-76-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR