Erbium ≥99.9% (trace rare earth metals basis), foil, Thickness: 0.62 mm

Supplier: BeanTown Chemical

211340-25X25MM 211340-50X50MM
BT211340-25X25EA 188 USD
BT211340-25X25 BT211340-50X50
Erbium ≥99.9% (trace rare earth metals basis), foil, Thickness: 0.62 mm
Erbium

CAS: 7440-52-0; EC No: 231-160-1; MDL No: MFCD00010987 Solid, 3.5g/25x25mm; Molecular Formula: Er; MW: 167.26 Melting Point: 1529°; Boiling Point: 2868° Density (g/mL): 9.062


  • Solid, 3.5g/25x25mm

Formula: Er
MW: 167.26
Boiling Pt: 2868 °C
Melting Pt: 1529 °C
Density: 9.062 g/mL
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR