Platinum iridium alloy (90:10 w%) ≥99.9% (trace metals basis), wire Ø 0.25 mm, annealed

Supplier: BeanTown Chemical

211335-10CM 211335-50CM
BT211335-10CMEA 105.55 USD
BT211335-10CM BT211335-50CM
Platinum iridium alloy (90:10 w%) ≥99.9% (trace metals basis), wire Ø 0.25 mm, annealed
Platinum iridium alloy (90:10 w%)

MDL No: MFCD02091683 Solid; Formula: Pt:Ir; 90:10 wt%,


  • Solid

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR