3,5-Dimethoxybenzyl alcohol ≥99%
Supplier: BeanTown Chemical
Synonyms:
(3,5-Dimethoxyphenyl)methanol
211270-1G
211270-25G
211270-5G
BT211270-1GEA
27
USD
BT211270-1G
BT211270-25G
BT211270-5G
3,5-Dimethoxybenzyl alcohol ≥99%
3,5-Dimethoxybenzyl alcohol
CAS: 705-76-0; EC No: 211-888-6; MDL No: MFCD00004641
Crystalline/Solid; Molecular Formula: C9H12O3; MW: 168.19
Melting Point: 43-46°; Flash point: 110°C (230°F)
- Crystalline/Solid
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol Melting Pt: 43…46 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00004641 CAS Number: 705-76-0 EINECS: 211-888-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...