Palladium(II) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate

211225-1G 211225-5G
BT211225-1GEA 144.63 USD
BT211225-1G BT211225-5G
Palladium(II) acetylacetonate
Palladium(II) acetylacetonate

CAS: 14024-61-4; EC No: 237-859-8; MDL No: MFCD00000025 Powder; Linear Formula: Pd(C5H7O2)2; MW: 304.64 Melting Point: 200-251° (decomposes); Boiling Point: 300°


  • Powder

Formula: C₁₀H₁₄O₄Pd
MW: 304.64 g/mol
Boiling Pt: 300 °C
Melting Pt: 200…251 °C (dec.)
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR