Iridium(III) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Iridium(III) 2,4-acetylacetonate, Tris(2,4-pentanedionato-O,O')iridium (III), Iridium (III) tris(2,4-pentanedionato-O,O'), Iridium (III) 2,4-pentanedionate

211215-1G 211215-250MG 211215-5G
BT211215-1GEA 234 USD
BT211215-1G BT211215-250MG BT211215-5G
Iridium(III) acetylacetonate
Iridium(III) acetylacetonate

CAS: 15635-87-7; EC No: 239-711-8; MDL No: MFCD00015353 Crystalline; Molecular Formula: C15H21IrO6; MW: 489.53 Melting Point: 269-271°


  • Crystalline

Formula: C₁₅H₂₁IrO₆
MW: 489.55
Melting Pt: 269…271 °C
MDL Number: MFCD00015353
CAS Number: 15635-87-7
EINECS: 239-711-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR