Indium(III) acetate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
211210-10G
211210-50G
BT211210-10GEA
106.37
USD
BT211210-10G
BT211210-50G
Indium(III) acetate ≥99.99% (trace metals basis)
Indium(III) acetate
CAS: 25114-58-3; MDL No: MFCD00054347
Powder; Linear Formula: In(OOCCH3)3; MW: 291.96
Melting Point: 270° (decomposes)
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₂H₄InO₂ MW: 174.87 g/mol Melting Pt: 270 °C (dec.) |
MDL Number:
MFCD00054347 CAS Number: 25114-58-3 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...