Trimethylammonium chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethylamine hydrochloride
211195-250G
211195-50G
BT211195-250GEA
50
USD
BT211195-250G
BT211195-50G
Trimethylammonium chloride ≥98%
Trimethylammonium chloride
CAS: 593-81-7; EC No: 209-810-0; MDL No: MFCD00012478; RTECS: YH2700000
Crystalline/Powder; Linear Formula: (CH3)3N·HCl; MW: 95.57
Melting Point: 283-284° (decomposes)
Density (g/mL): 1.04
- RTECS: YH2700000
- Crystalline/Powder
Formula:
C₃H₁₀ClN MW: 95.57 g/mol Melting Pt: 283…284 °C (dec.) Density: 1.04 g/mL |
MDL Number:
MFCD00012478 CAS Number: 593-81-7 EINECS: 209-810-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...