2'-Methylacetanilide ≥98%

Supplier: BeanTown Chemical

Synonyms: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide

211185-100G 211185-25G 211185-500G
BT211185-100GEA 85.27 USD
BT211185-100G BT211185-25G BT211185-500G
2'-Methylacetanilide ≥98%
2'-Methylacetanilide

CAS: 120-66-1; EC No: 204-414-4; MDL No: MFCD00014961; RTECS: AN2900000 Powder; Molecular Formula: C9H11NO; MW: 149.19 Density (g/mL): 1.16


  • RTECS: AN2900000
  • Powder

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Density: 1.16 g/mL
MDL Number: MFCD00014961
CAS Number: 120-66-1
EINECS: 204-414-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR