1-Tetradecanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol

211155-100G 211155-2.5KG 211155-500G
BT211155-100GEA 28.9 USD
BT211155-100G BT211155-2.5KG BT211155-500G
1-Tetradecanol ≥97%
1-Tetradecanol

CAS: 112-72-1; EC No: 204-000-3; MDL No: MFCD00004757; RTECS: XB8655000 Solid; Linear Formula: CH3(CH2)12CH2OH; Molecular Formula: C14H30O; MW: 214.39 Melting Point: 35-39°; Boiling Point: 289°; Flash point: 148°C (298°F) Density (g/mL): 0.823


  • RTECS: XB8655000
  • Solid

Formula: H₃C(CH₂)₁₂CH₂OH
MW: 214.39 g/mol
Boiling Pt: 289
Melting Pt: 35…39 °C
Density: 0.823 g/mL
Flash Pt: 148 °C (298 °F)
MDL Number: MFCD00004757
CAS Number: 112-72-1
EINECS: 204-000-3
Merck Index: 13,06361

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR