4-(Methylsulfonyl)phenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

211130-1G 211130-250MG 211130-5G
BT211130-1GEA 105 USD
BT211130-1G BT211130-250MG BT211130-5G
4-(Methylsulfonyl)phenylboronic acid ≥98%
4-(Methylsulphonyl)phenylboronic acid

CAS: 149104-88-1; MDL No: MFCD01630820 Solid; Molecular Formula: C7H9BO4S; MW: 200.02 Melting Point: 289-293°


  • Solid

Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Melting Pt: 289…293 °C
MDL Number: MFCD01630820
CAS Number: 149104-88-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR