Iodobenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Phenyliodide
211100-100G
211100-500G
BT211100-100GEA
87
USD
BT211100-100G
BT211100-500G
Iodobenzene ≥98%
Iodobenzene
CAS: 591-50-4; EC No: 209-719-6; MDL No: MFCD00001029; RTECS: DA3390000
Liquid; Molecular Formula: C6H5I; MW: 204.01
Melting Point: -29°; Boiling Point: 188°; Flash point: 74°C (165°F)
Density (g/mL): 1.823; Refractive Index: 1.62
Light Sensitive
- RTECS: DA3390000
- Refractive index: 1.62
- Light Sensitive
- Liquid
Formula:
C₆H₅I MW: 204.01 g/mol Boiling Pt: 188 °C Melting Pt: –29 °C Density: 1.823 g/mL Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00001029 CAS Number: 591-50-4 EINECS: 209-719-6 |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...