trans-N-Boc-4-cyano-L-proline ≥97%

Supplier: BeanTown Chemical

211095-1G 211095-250MG
BT211095-1GEA 1161 USD
BT211095-1G BT211095-250MG
trans-N-Boc-4-cyano-L-proline ≥97%
trans-N-Boc-4-cyano-L-proline

CAS: 273221-94-6; MDL No: MFCD01860670 Solid; Molecular Formula: C11H16N2O4 ; MW: 240.26


  • Solid

Formula: C₁₁H₁₆N₂O₄
MDL Number: MFCD01860670
CAS Number: 273221-94-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR