6-Bromo-1-hexanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 6-Bromohexan-1-ol, Hexamethylene bromohydrin, 1-Bromo-6-hydroxyhexane

211090-1G 211090-25G 211090-5G
BT211090-1GEA 34 USD
BT211090-1G BT211090-25G BT211090-5G
6-Bromo-1-hexanol ≥97%
6-Bromo-1-hexanol

CAS: 4286-55-9; MDL No: MFCD00002983 Liquid; Linear Formula: BrCH2(CH2)4CH2OH; Molecular Formula: C6H13BrO; MW: 181.07 Boiling Point: 105-106°/5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.384; Refractive Index: 1.482


  • Refractive index: 1.482
  • Liquid

Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 105…106 °C/5 mmHg
Density: 1.384 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00002983
CAS Number: 4286-55-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR