9-Bromoanthracene ≥96%

Supplier: BeanTown Chemical

211045-1G 211045-5G
BT211045-1GEA 22 USD
BT211045-1G BT211045-5G
9-Bromoanthracene ≥96%
9-Bromoanthracene

CAS: 1564-64-3; EC No: 216-359-3; MDL No: MFCD00001243 Crystalline; Molecular Formula: C14H9Br; MW: 257.13 Melting Point: 97-100°


  • Crystalline

Formula: C₁₄H₉Br
MW: 257.13 g/mol
Melting Pt: 97…100 °C
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR