Calcium carbonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Marble
211035-2KG
211035-500G
BT211035-2KGEA
112.51
USD
BT211035-2KG
BT211035-500G
Calcium carbonate ≥98%
Calcium carbonate
CAS: 471-34-1; EC No: 207-439-9; MDL No: MFCD00010906; RTECS: FF9335000
Powder; Linear Formula: CaCO3; MW: 100.09
Density (g/mL): 2.93
- RTECS: FF9335000
- Powder
Formula:
CaCO₃ MW: 100.09 g/mol Density: 2.93 g/mL |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...