Isopropyl myristate ≥98%
Supplier: BeanTown Chemical
CAS: 110-27-0; EC No: 203-751-4; MDL No: MFCD00008982
Liquid; Linear Formula: CH3(CH2)12CO2CH(CH3)2; Molecular Formula: C17H34O2; MW: 270.46
Melting Point: -3°; Boiling Point: 193°/20 mmHg; Flash point: 152°C (306°F)
Density (g/mL): 0.85
- Liquid
Formula:
C₁₇H₃₄O₂ Boiling Pt: 193 °C/20 mmHg Melting Pt: –3 °C Density: 0.85 g/mL Flash Pt: 152 °C (306 °F) |
MDL Number:
MFCD00008982 CAS Number: 110-27-0 EINECS: 203-751-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...