5'-Bromo-2'-hydroxyacetophenone ≥98%
Supplier: BeanTown Chemical
145980-10G
145980-50G
BT145980-10GEA
43
USD
BT145980-10G
BT145980-50G
5'-Bromo-2'-hydroxyacetophenone ≥98%
5'-Bromo-2'-hydroxyacetophenone
CAS: 1450-75-5; MDL No: MFCD00191850; RTECS: KM5556350
Crystalline/Powder; Molecular Formula: C8H7BrO2; MW: 215.04
Melting Point: 58-61°
- RTECS: KM5556350
- Crystalline/Powder
Formula:
C₈H₇BrO₂ Melting Pt: 58…61 °C |
MDL Number:
MFCD00191850 CAS Number: 1450-75-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...